Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
btbd2a
Entrez Gene ID
565970
Gene Name
BTB (POZ) domain containing 2a
Gene Aliases
btb2l, btbd2, si:dkey-110c1.5, wz8656
Chromosome Location
Chr.22: 20159064 - 20166697 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045092.1 NP_001038557.1 3 - 4 972 103
XM_021469350.1 XP_021325025.1 3 - 4 872 103
XM_021469351.1 XP_021325026.1 2 - 3 843 103
XM_021469352.1 XP_021325027.1 3 - 4 500 103
GenBank mRNA BC163262.1 - 3 - 4 849 103
DQ241789.1 - 3 - 4 972 103

View Full Product Details