Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
flr
Entrez Gene ID
797829
Gene Name
fleer
Gene Aliases
ttc30, ttc30a
Chromosome Location
Chr.3: 28831470 - 28850898 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104649.2 NP_001098119.2 12 - 13 1236 110
XM_021471707.1 XP_021327382.1 11 - 12 1209 110
XM_021471709.1 XP_021327384.1 10 - 11 999 110
XM_021471716.1 XP_021327391.1 12 - 13 1298 110
GenBank mRNA BC163912.1 - 12 - 13 1258 110
EF653429.1 - 12 - 13 1318 110

View Full Product Details