Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
shhb
Entrez Gene ID
30444
Gene Name
sonic hedgehog signaling molecule b
Gene Aliases
twh, twhh
Chromosome Location
Chr.2: 30050547 - 30055381 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131199.2 NP_571274.2 1 - 2 476 97
GenBank mRNA BC078321.1 - 1 - 2 476 97
KM655588.1 - 1 - 2 310 97
U30710.1 - 1 - 2 310 97

View Full Product Details