Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cry2
Entrez Gene ID
83774
Gene Name
cryptochrome circadian regulator 2
Gene Aliases
cry3, fe16d07, wu:fe16d07, zcry3
Chromosome Location
Chr.25: 13847458 - 13871118 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131786.2 NP_571861.2 7 - 8 1395 82
GenBank mRNA AB042252.1 - 7 - 8 1284 82
BC046088.1 - 7 - 8 1395 82

View Full Product Details