Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
brf1b
Entrez Gene ID
334316
Gene Name
BRF1 RNA polymerase III transcription initiation factor subunit b
Gene Aliases
fi40c10, wu:fi40c10, zgc:63497
Chromosome Location
Chr.20: 20901791 - 21016899 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199889.1 NP_956183.1 12 - 13 1565 85
GenBank mRNA BC060523.1 - 12 - 13 1565 85

View Full Product Details