Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gorasp2
Entrez Gene ID
393676
Gene Name
golgi reassembly stacking protein 2
Gene Aliases
zgc:73072
Chromosome Location
Chr.6: 3864016 - 3875115 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200703.2 NP_956997.1 2 - 3 261 87
XM_021476811.1 XP_021332486.1 2 - 3 285 87
GenBank mRNA AY398354.1 - 2 - 3 261 87
BC059447.1 - 2 - 3 222 87

View Full Product Details