Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rab22a
Entrez Gene ID
100001531
Gene Name
RAB22A, member RAS oncogene family
Gene Aliases
im:7136651, zgc:101631
Chromosome Location
Chr.6: 10382 - 35069 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205719.2 NP_991282.2 3 - 4 441 130
GenBank mRNA AY398404.1 - 3 - 4 478 130
BC085393.1 - 3 - 4 441 130
BC165262.1 - 3 - 4 233 130

View Full Product Details