Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bambib
Entrez Gene ID
431736
Gene Name
BMP and activin membrane-bound inhibitor homolog (Xenopus laevis) b
Gene Aliases
zgc:91871
Chromosome Location
Chr.2: 4402765 - 4408829 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002189.1 NP_001002189.1 1 - 2 358 92
GenBank mRNA BC072715.1 - 1 - 2 358 92
BC165294.1 - 1 - 2 107 92

View Full Product Details