Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
zdhhc15a
Entrez Gene ID
449833
Gene Name
zinc finger DHHC-type palmitoyltransferase 15a
Gene Aliases
zgc:103780
Chromosome Location
Chr.5: 300060 - 324315 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006003.1 NP_001006003.1 1 - 2 145 84
XM_021476124.1 XP_021331799.1 1 - 2 148 84
GenBank mRNA BC083491.1 - 1 - 2 145 84

View Full Product Details