Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tmpob
Entrez Gene ID
553624
Gene Name
thymopoietin b
Gene Aliases
wu:fa17b10, wu:fi69e10, zgc:110551
Chromosome Location
Chr.25: 3294374 - 3302666 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020598.1 NP_001018434.1 3 - 4 690 97
GenBank mRNA BC095307.1 - 3 - 4 690 97

View Full Product Details