Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
npr1a
Entrez Gene ID
560653
Gene Name
natriuretic peptide receptor 1a
Gene Aliases
sGC, si:dkey-27c15.1
Chromosome Location
Chr.19: 26423073 - 26627313 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044937.2 NP_001038402.1 3 - 4 1052 143
XM_005170050.4 XP_005170107.1 4 - 5 1732 143

View Full Product Details