Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mixl1
Entrez Gene ID
30115
Gene Name
Mix paired-like homeobox
Gene Aliases
bon, bon/mixer, mix, mix-like
Chromosome Location
Chr.20: 43715157 - 43723765 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130940.3 NP_571015.3 2 - 3 344 58
GenBank mRNA AF121771.1 - 2 - 3 447 58
BC097091.1 - 2 - 3 346 58

View Full Product Details