Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
add3a
Entrez Gene ID
556762
Gene Name
adducin 3 (gamma) a
Gene Aliases
add3, wu:fb38g03, wu:fc16e06, zgc:55560, zgc:55992, zgc:77226
Chromosome Location
Chr.22: 30047220 - 30272394 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199663.1 NP_955957.1 5 - 6 509 66
XM_005161804.4 XP_005161861.1 2 - 3 515 66
XM_005161805.4 XP_005161862.1 2 - 3 515 66
XM_005161807.4 XP_005161864.1 2 - 3 514 66
XM_017353146.2 XP_017208635.1 2 - 3 516 66
XM_017353147.2 XP_017208636.1 3 - 4 386 66
XM_017353148.2 XP_017208637.1 2 - 3 734 66
XM_017353149.2 XP_017208638.1 2 - 3 516 66
XM_017353150.2 XP_017208639.1 2 - 3 515 66
XM_021469329.1 XP_021325004.1 4 - 5 483 66
XM_021469330.1 XP_021325005.1 2 - 3 244 66
XM_021469331.1 XP_021325006.1 4 - 5 482 66
GenBank mRNA AY391406.1 - 1 - 2 223 66
BC045384.1 - 2 - 3 413 66
BC050486.1 - 5 - 6 509 66
BC065631.1 - 2 - 3 475 66
BC165643.1 - 1 - 2 230 66

View Full Product Details