The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_200183.1 | NP_956477.1 | 5 - 6 | 1249 | 61 |
GenBank mRNA | BC045490.1 | - | 5 - 6 | 1249 | 61 |