Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
znhit1
Entrez Gene ID
407699
Gene Name
zinc finger, HIT-type containing 1
Gene Aliases
zgc:112524
Chromosome Location
Chr.5: 38604877 - 38611407 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017401.1 NP_001017401.1 4 - 5 479 69
GenBank mRNA BC067648.1 - 4 - 5 433 69
BC093363.1 - 4 - 5 479 69

View Full Product Details