Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
apobb.1
Entrez Gene ID
321166
Gene Name
apolipoprotein Bb, tandem duplicate 1
Gene Aliases
apobb, apobl, cb235, hm:zeh1207, sb:cb235, si:ch211-219i10.1, wu:fb39c11, wu:fc51e12, zeh1207
Chromosome Location
Chr.20: 31269702 - 31284894 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030062.1 NP_001025233.1 21 - 22 3527 120
XM_005160629.3 XP_005160686.1 22 - 23 3630 120

View Full Product Details