Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nanog
Entrez Gene ID
792333
Gene Name
nanog homeobox
Gene Aliases
fd20a07, hacp, wu:fd19e04, wu:fd20a07, zgc:193933
Chromosome Location
Chr.24: 12836079 - 12841158 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098392.1 NP_001091862.1 3 - 4 1006 78
GenBank mRNA BC162315.1 - 3 - 4 940 78
BC162318.1 - 3 - 4 940 78
EF550998.1 - 3 - 4 1006 78
JN615142.1 - 3 - 4 971 78
JN809237.1 - 3 - 4 833 78

View Full Product Details