Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lctlb
Entrez Gene ID
564250
Gene Name
lactase-like b
Gene Aliases
si:rp71-14b1.1
Chromosome Location
Chr.18: 19467305 - 19484742 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100014.2 NP_001093484.1 12 - 13 1566 66
XM_005159111.3 XP_005159168.1 13 - 14 1785 66

View Full Product Details