Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pax6b
Entrez Gene ID
60639
Gene Name
paired box 6b
Gene Aliases
Pax6.2, cb566, fc09c10, id:ibd5095, wu:fc09c10, wu:fc33g08
Chromosome Location
Chr.7: 15871376 - 15897386 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131641.1 NP_571716.1 4 - 5 457 105
XM_005168823.4 XP_005168880.1 2 - 3 420 105
XM_009302973.3 XP_009301248.1 4 - 5 484 105
XM_021477603.1 XP_021333278.1 5 - 6 495 105
GenBank mRNA AF061252.1 - 4 - 5 457 105
BC076068.1 - 4 - 5 478 105

View Full Product Details