Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
glra3
Entrez Gene ID
192124
Gene Name
glycine receptor, alpha 3
Gene Aliases
glyRalpha3
Chromosome Location
Chr.1: 38416268 - 38538252 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152965.1 NP_694497.1 8 - 9 1297 80
GenBank mRNA AJ308516.1 - 8 - 9 1297 80
BC163529.1 - 8 - 9 1197 80

View Full Product Details