Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
szl
Entrez Gene ID
353294
Gene Name
sizzled
Gene Aliases
sfrp, slz
Chromosome Location
Chr.25: 371412 - 375892 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181663.1 NP_858049.1 3 - 4 549 84
GenBank mRNA AB093587.1 - 3 - 4 549 84
AY189978.2 - 3 - 4 572 84
BC094990.1 - 3 - 4 562 84

View Full Product Details