Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
setd6
Entrez Gene ID
322348
Gene Name
SET domain containing 6, protein lysine methyltransferase
Gene Aliases
wu:fb58f11, zgc:55627
Chromosome Location
Chr.25: 17349265 - 17357132 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199600.1 NP_955894.1 4 - 5 561 120
GenBank mRNA BC044440.1 - 4 - 5 577 120

View Full Product Details