Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
setd6
Entrez Gene ID
322348
Gene Name
SET domain containing 6, protein lysine methyltransferase
Gene Aliases
wu:fb58f11, zgc:55627
Chromosome Location
Chr.25: 17349265 - 17357132 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199600.1 NP_955894.1 7 - 8 1075 64
GenBank mRNA BC044440.1 - 7 - 8 1091 64

View Full Product Details