Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hif1al
Entrez Gene ID
393376
Gene Name
hypoxia inducible factor 1 subunit alpha, like
Gene Aliases
Hif-3alpha, zgc:63965
Chromosome Location
Chr.15: 25503955 - 25527622 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200405.1 NP_956699.1 8 - 9 1061 86
XM_005157571.4 XP_005157628.2 8 - 9 1131 86
XM_005157572.4 XP_005157629.2 8 - 9 1130 86
XM_021481358.1 XP_021337033.1 6 - 7 826 86
GenBank mRNA AY330295.1 - 8 - 9 1089 86
BC054582.1 - 8 - 9 1061 86
GQ253368.1 - 8 - 9 1095 86

View Full Product Details