Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lctla
Entrez Gene ID
437008
Gene Name
lactase-like a
Gene Aliases
zgc:101102
Chromosome Location
Chr.7: 192785 - 201827 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002735.2 NP_001002735.1 8 - 9 838 119
GenBank mRNA BC076422.1 - 8 - 9 842 119

View Full Product Details