Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tspan7b
Entrez Gene ID
449539
Gene Name
tetraspanin 7b
Gene Aliases
tm4sf2b, wu:fb53h11, zgc:92829
Chromosome Location
Chr.22: 11440705 - 11493236 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130818.1 NP_001124290.1 5 - 6 648 83
GenBank mRNA BC081682.1 - 5 - 6 599 83

View Full Product Details