Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
atp6v1c1b
Entrez Gene ID
449655
Gene Name
ATPase H+ transporting V1 subunit C1b
Gene Aliases
atp6v1c1l, si:zc215i13.2, zgc:92684
Chromosome Location
Chr.19: 84560 - 103412 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005772.2 NP_001005772.2 10 - 11 1100 79
GenBank mRNA BC083532.1 - 10 - 11 1004 79

View Full Product Details