Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cntn1b
Entrez Gene ID
541474
Gene Name
contactin 1b
Gene Aliases
si:dkey-240a12.2
Chromosome Location
Chr.4: 13733838 - 13756711 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014814.1 NP_001014814.1 4 - 5 459 96
GenBank mRNA AY301281.1 - 4 - 5 459 96

View Full Product Details