Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fstl3
Entrez Gene ID
557352
Gene Name
follistatin-like 3 (secreted glycoprotein)
Gene Aliases
si:dkey-28h18.3, zgc:114170
Chromosome Location
Chr.22: 18884872 - 18893390 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030111.2 NP_001025282.2 2 - 3 354 74
GenBank mRNA BC097220.1 - 2 - 3 329 74
BC134879.1 - 2 - 3 354 74

View Full Product Details