Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
btd
Entrez Gene ID
555376
Gene Name
biotinidase
Gene Aliases
zgc:153333
Chromosome Location
Chr.19: 1999935 - 2004248 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045844.1 NP_001039309.1 2 - 3 498 66
GenBank mRNA BC122246.1 - 2 - 3 498 66

View Full Product Details