Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
txlnbb
Entrez Gene ID
564166
Gene Name
taxilin beta b
Gene Aliases
zgc:162968
Chromosome Location
Chr.17: 6358160 - 6382392 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083553.2 NP_001077022.3 10 - 11 1214 80
GenBank mRNA BC135082.1 - 10 - 11 80

View Full Product Details