Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
txlnbb
Entrez Gene ID
564166
Gene Name
taxilin beta b
Gene Aliases
zgc:162968
Chromosome Location
Chr.17: 6358160 - 6382392 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083553.2 NP_001077022.3 9 - 10 1121 82
NM_001328168.1 NP_001315097.1 9 - 10 1124 82
XM_005158569.3 XP_005158626.1 9 - 10 1116 82
GenBank mRNA BC135082.1 - 9 - 10 1120 82

View Full Product Details