Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rims3
Entrez Gene ID
563414
Gene Name
regulating synaptic membrane exocytosis 3
Gene Aliases
si:ch211-239d6.1
Chromosome Location
Chr.19: 25162602 - 25301565 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100009.2 NP_001093479.1 4 - 5 561 64
XM_005170052.4 XP_005170109.1 5 - 6 839 64

View Full Product Details