Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gprc5c
Entrez Gene ID
563405
Gene Name
G protein-coupled receptor, class C, group 5, member C
Gene Aliases
zgc:171736
Chromosome Location
Chr.12: 36023230 - 36045336 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109713.1 NP_001103183.1 3 - 4 1468 68
GenBank mRNA BC151953.1 - 3 - 4 1468 68

View Full Product Details