Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
birc2
Entrez Gene ID
373107
Gene Name
baculoviral IAP repeat containing 2
Gene Aliases
IAP1, birc3
Chromosome Location
Chr.21: 22833892 - 22851099 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194395.3 NP_919376.1 5 - 6 1842 85
XM_005161156.4 XP_005161213.1 5 - 6 1910 85
XM_009295293.3 XP_009293568.1 6 - 7 2162 85
XM_017352755.2 XP_017208244.1 5 - 6 1865 85
GenBank mRNA AF442500.1 - 4 - 5 1190 85
AY247786.1 - 5 - 6 1807 85
BC115241.1 - 4 - 5 1237 85
BC124076.1 - 5 - 6 1843 85
BC165235.1 - 4 - 5 1186 85

View Full Product Details