Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
flot1b
Entrez Gene ID
245697
Gene Name
flotillin 1b
Gene Aliases
cb2, cb227, reggie-2b, sb:cb2, sb:cb227, wu:fb53d05
Chromosome Location
Chr.16: 10287588 - 10316359 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201456.1 NP_958864.1 10 - 11 1061 63
GenBank mRNA AY398349.1 - 10 - 11 1061 63

View Full Product Details