Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cth
Entrez Gene ID
322055
Gene Name
cystathionase (cystathionine gamma-lyase)
Gene Aliases
fb58d08, wu:fb48e03, wu:fb58d08, zgc:66001, zgc:85785
Chromosome Location
Chr.6: 37409558 - 37422947 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212604.3 NP_997769.3 7 - 8 894 82
GenBank mRNA BC056538.1 - 7 - 8 780 82
EH493556.1 - 7 - 8 831 82

View Full Product Details