Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
plppr1
Entrez Gene ID
415222
Gene Name
phospholipid phosphatase related 1
Gene Aliases
lppr1, sb:cb978, zgc:86759
Chromosome Location
Chr.10: 4240816 - 4264026 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002132.1 NP_001002132.1 - 826 83
GenBank mRNA BC071430.1 - - 826 83

View Full Product Details