Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ctdnep1b
Entrez Gene ID
492799
Gene Name
CTD nuclear envelope phosphatase 1b
Gene Aliases
ctdnep1l, dullardl, zgc:101656
Chromosome Location
Chr.7: 69449858 - 69467638 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007440.2 NP_001007441.2 3 - 4 770 139
GenBank mRNA BC085403.1 - 3 - 4 139

View Full Product Details