Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dpysl5a
Entrez Gene ID
324416
Gene Name
dihydropyrimidinase like 5a
Gene Aliases
CRMP-5a, DPYSL5, crmp5.1, fc28b06, im:7146614, wu:fc28b06
Chromosome Location
Chr.17: 36899602 - 36936649 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018149.1 NP_001018159.1 10 - 11 1386 68
GenBank mRNA AY987371.1 - 10 - 11 1386 68
BC124748.1 - 10 - 11 1337 68
BC165313.1 - 9 - 10 1260 68

View Full Product Details