Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
crcp
Entrez Gene ID
751656
Gene Name
calcitonin gene-related peptide-receptor component protein
Gene Aliases
rcp9, zgc:153737
Chromosome Location
Chr.21: 31244278 - 31252167 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045373.2 NP_001038838.2 5 - 6 361 75
XM_017352825.2 XP_017208314.1 5 - 6 385 75
XM_021468909.1 XP_021324584.1 5 - 6 336 75
GenBank mRNA BC164588.1 - 4 - 5 328 75
EH585180.1 - 5 - 6 353 75

Gene Symbol
LOC110438496
Entrez Gene ID
110438496
Gene Name
DNA-directed RNA polymerase III subunit RPC9-like
Gene Aliases
-
Chromosome Location
Chr.Unknown: 1822 - 6175 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_021471041.1 XP_021326716.1 2 - 3 209 75

View Full Product Details