Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vsx1
Entrez Gene ID
30598
Gene Name
visual system homeobox 1 homolog, chx10-like
Gene Aliases
-
Chromosome Location
Chr.17: 123985 - 126655 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131333.1 NP_571408.1 1 - 2 416 64
XM_005158663.4 XP_005158720.1 1 - 2 439 64
GenBank mRNA AF025348.2 - 1 - 2 416 64
BC059574.1 - 1 - 2 408 64

View Full Product Details