Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ncln
Entrez Gene ID
336844
Gene Name
nicalin
Gene Aliases
ncl1, wu:fb07e09, wu:fk52a06, zgc:77236
Chromosome Location
Chr.22: 5233396 - 5252015 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207057.1 NP_996940.1 14 - 15 1759 102
XM_005161589.4 XP_005161646.1 14 - 15 1759 102
XM_005161590.4 XP_005161647.1 14 - 15 1717 102
XM_005161591.4 XP_005161648.1 14 - 15 1714 102
GenBank mRNA BC056523.1 - 14 - 15 1749 102
BC066393.1 - 14 - 15 1759 102

View Full Product Details