Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sec22ba
Entrez Gene ID
114464
Gene Name
SEC22 homolog B, vesicle trafficking protein a
Gene Aliases
sec22l1, sec22l1a, wu:fb53h12, wu:fc15g01, zgc:56278
Chromosome Location
Chr.2: 9973687 - 9983661 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213384.1 NP_998549.1 1 - 2 265 82
GenBank mRNA BC046043.1 - 1 - 2 265 82

View Full Product Details