Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpp40
Entrez Gene ID
415238
Gene Name
ribonuclease P/MRP 40 subunit
Gene Aliases
rpp40l, zgc:86704
Chromosome Location
Chr.2: 23613145 - 23620219 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002148.1 NP_001002148.1 4 - 5 499 91
GenBank mRNA BC071381.1 - 4 - 5 499 91

View Full Product Details