Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hormad1
Entrez Gene ID
436630
Gene Name
HORMA domain containing 1
Gene Aliases
zgc:92129
Chromosome Location
Chr.16: 29551026 - 29557612 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002357.1 NP_001002357.1 14 - 15 1164 59
XM_021466675.1 XP_021322350.1 12 - 13 1771 59
GenBank mRNA BC075898.1 - 14 - 15 1164 59

View Full Product Details