Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gata2b
Entrez Gene ID
436962
Gene Name
GATA binding protein 2b
Gene Aliases
zgc:91840
Chromosome Location
Chr.6: 40794005 - 40803812 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002689.1 NP_001002689.1 2 - 3 340 118
XM_009302543.3 XP_009300818.1 1 - 2 741 118
GenBank mRNA BC076462.1 - 2 - 3 340 118
BC165802.1 - 1 - 2 282 118

View Full Product Details