Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vash2
Entrez Gene ID
562668
Gene Name
vasohibin 2
Gene Aliases
si:ch211-147d7.1
Chromosome Location
Chr.20: 37866185 - 37916517 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025523.1 NP_001020694.1 1 - 2 321 81
XM_005160740.4 XP_005160797.1 1 - 2 822 81

View Full Product Details