Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sat1a.1
Entrez Gene ID
565700
Gene Name
spermidine/spermine N1-acetyltransferase 1a, duplicate 1
Gene Aliases
sat1a, zgc:114142
Chromosome Location
Chr.5: 4332220 - 4342795 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030199.1 NP_001025370.1 2 - 3 287 116
GenBank mRNA BC097197.1 - 2 - 3 287 116
BC171337.1 - 2 - 3 269 116
BC171340.1 - 2 - 3 269 116

View Full Product Details