Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
eefsec
Entrez Gene ID
641497
Gene Name
eukaryotic elongation factor, selenocysteine-tRNA-specific
Gene Aliases
zgc:123265
Chromosome Location
Chr.6: 40822587 - 40832246 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037436.1 NP_001032513.1 5 - 6 1599 74
GenBank mRNA BC109397.1 - 5 - 6 1599 74

View Full Product Details